Polyethylene [ST] Stoppers, Chemglass
Supplier: Chemglass
CG-3021-04 CG-3021-01 CG-3021-02 CG-3021-03
33501-438PK 50.65 CAD
33501-438 33501-430 33501-432 33501-434
Polyethylene [ST] Stoppers, Chemglass
Stoppers PE/LDPE/HDPE Stoppers
Inexpensive, tapered stoppers provide a tight, leakproof fit while the top ring permits the stopper to be removed easily
Stoppers are made of yellow polyethylene.