PTFE Key-Head [ST] Stoppers, Flask Length Joint, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
41901R-32 41901R-38 41901R-9 41901R-13 41901R-16 41901R-19 41901R-22
16358-236CS 510.5 CAD
16358-236 16358-238 89000-830 89000-832 89000-834 89000-836 89000-838
PTFE Key-Head [ST] Stoppers, Flask Length Joint, Kimble Chase
Stoppers PE/LDPE/HDPE Stoppers
Stoppers are for use with flasks, mixing cylinders, separatory funnels, and more
Designed from ASTM Specification E675.
Color-coded handles indicate stopper size. Stoppers size 9 through 22 are solid. Sizes 32 and 38 are hollow.