Socorex™ Dosys™ 163/173 Classic Self-Refilling Syringes, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
W852006 W852008 W852010 W852012 W853008 W853010 W853012 W853014 W853016 W853018 W853020
89044-872EA 651.71 CAD
89044-872 89044-874 89044-876 89044-878 89044-896 89044-898 89044-900 89044-902 89044-904 89044-906 89044-908
Socorex™ Dosys™ 163/173 Classic Self-Refilling Syringes, DWK Life Sciences
Syringes General-Purpose Syringes
Self-refilling, automatic syringes feature a spring-loaded plunger that allows repetitive delivery of preset aliquots from a single reservoir
Handle has been designed to provide fatigue-free operation over an extended period of time. Integrated valve system with corrosion-resistant springs and stainless steel valve balls enhances repetitive accuracy. Syringes also feature an interchangeable ground glass barrel with corrosion-resistant stainless steel plunger, Viton® washers, and Luer lock nozzle. Autoclavable.
Ordering information: One-year manufacturer's warranty. Each syringe includes a 1m (393/8") silicone feed tube, sinker, PVC protection sleeve, aspiration and vent cannulas, set of spare valves, and operating instructions. For accessories and replacement parts, see 89044-552 series.