KONTES® Solid Pennyhead [ST] Stoppers, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
850100-0016 850100-0019 850100-0022 850100-0008 850100-0009 850100-0013 850100-0027 850100-0038
KT850100-0016EA 51.88 CAD
KT850100-0016 KT850100-0019 KT850100-0022 KT850100-0008 KT850100-0009 KT850100-0013 KT850100-0027 KT850100-0038
KONTES® Solid Pennyhead [ST] Stoppers, Kimble Chase
Stoppers Glass Stoppers
Glass stoppers for use with flasks, mixing cylinders, separatory funnels, and more
Meet ASTM Specification E694.
All stoppers are solid.