PCR 8-Strip Tubes and Caps
Supplier: NEST SCIENTIFIC
403112 403122 403132 403002 403012
77508-054CS 628.29 CAD
77508-054 77508-056 77508-058 77508-044 77508-046
PCR 8-Strip Tubes and Caps
Tubes PCR Tubes
Transparent and white PCR strip tubes to the general PCR and qPCR reactions.
  • Made from USP grade VI polypropylene
  • Non-pyrogenic, Free of DNase and RNase, human DNA and PCR inhibitor
  • Seemed animal free
  • Autoclavable (121 ℃, 20 min)
  • White strip tubes increase the fluorescent signal strength of real-time PCR
  • Stored at room temperature
Low profile 0.1 ml tubes which are designed to minimize the condensate contamination, particularly for the overnight experiment, and also reduce the evaporation of the reaction solution.
Universally applicable to theMore Product Information