Storage Bottles, Square, Sterile

Supplier: NEST SCIENTIFIC
333621 333624 334511 352511 352513 353511 353513 333001 333511 354113 354511 354513 355114 355314
77507-690CS 147.21 CAD
77507-690 77507-692 77507-696 77507-954 77507-956 77507-960 77507-962 77507-684 77507-686 77507-968 77507-970 77507-972 77507-976 77507-978
Storage Bottles, Square, Sterile
Bottles Cell Culture Bottles
NEST’s Square storage bottle is an ideal option of storage and transportation container for culture medium, serum, buffer, intermediates and reagents for biopharmaceutical and biotechnology companies, reagent R and D institutions, etc.

  • PET wall is light in weight and resistant to low temperature, while PETG wall is thick, durable, and resistant to fracture
  • High chemical durability, suitable for the storage of high purity reagent, standard particle materials and samples
  • High transparency, strong mechanical strength and shock resistance, and convenient for observation and transportation
  • Highly tolerant of low temperature, UV rays, and resistant to fracture
  • Smooth inner surface can minimize the residue on the wall
  • Sterile, DNase/RNase/Pyrogen free

Applications: Storage or sampling of active pharmaceutical ingredients and bulk intermediates, experiments or storage of buffers, culture solutions, intermediates and supernatants, etc, storage of specific PH-sensitive liquid.

Certifications: The two kinds of raw materials of the bottle, PET and PETG, are both in line with USP Class VI Class USP<661> biocompatibility requirements and ISO10993 Quality System.

Caution: Unfit for autoclave. Not a vessel to be heated.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR