Deepwell Plates 96-well, 500 µl
Supplier: Eppendorf
951031801 951032204 951031909 951032301 0030113543
CA89085-642CS 714.35 CAD
CA89085-642 CA89085-644 CA89085-646 CA89085-648 CA77395-524
Deepwell Plates 96-well, 500 µl
Microplates Deep Well Microplates
Eppendorf Deepwell plates 96-well, 500 µl offer efficient sample handling with features like OptiTrack® matrix for rapid identification and g-Safe® technology for stability during centrifugation. SafeCode variants provide secure three-level coding, including error correction for reliable identification, ensuring flexibility and accuracy in experimental workflows.
  • OptiTrack® matrix: up to 30% faster sample identification and fewer pipetting errors via high-contrast alphanumeric labeling
  • RecoverMax® well design: Optimized well geometry for maximum sample
More Product Information