Synthware Stopcock, High Vacuum, T-Bore Hollow Glass Plug, Kemtech America
Supplier: Kemtech America
S721240 S721754
60005-216EA 176.91 CAD
60005-216 60005-218
Synthware Stopcock, High Vacuum, T-Bore Hollow Glass Plug, Kemtech America
Stopcocks
High vacuum stopcock with T-bore hollow glass plug.
  • Heavy wall borosilicate construction
  • Hollow plug
  • T-type
Barrel and plug are precision ground with corresponding numbers on the barrel and plug. The lower vacuum chamber is evacuated via a separate 2mm plug hole, which is not in the line of flow.