Quantitative Fit Testing Respirator, 3M™
Supplier: Avantor
CA56222-920CS 184.51 CAD
CA56222-920 CA56222-922 CA56222-924
Quantitative Fit Testing Respirator, 3M™
Respirators Respiratory Systems
Developed exclusively for quantitative face-fit testing to ensure proper worker protection.
  • Not designed for respiratory protection
Features the 3M 6000 series facepiece, with a probe installed for testing convenience.
Packaging: Caution: All 3M™ products must be used in accordance with OSHA regulations and the user instructions, warnings, and limitations accompanying each product.
Caution: All 3M™ products must be used in accordance with OSHA regulations and the user instructions, warnings, and limitations accompanying each product.