Synthware Stopcock, High Vacuum, Offset Bore Hollow Glass Plug, Kemtech America
Supplier: Kemtech America
S701240 S701754 S701756
60005-204EA 160.64 CAD
60005-204 60005-206 60005-208
Synthware Stopcock, High Vacuum, Offset Bore Hollow Glass Plug, Kemtech America
Stopcocks
High vacuum stopcock with offset bore hollow glass plug.
  • Heavy wall borosilicate construction
  • Hollow plug
  • Offset
Barrel and plug are precision ground with corresponding numbers on the barrel and plug. The lower vacuum chamber is evacuated via a separate 2 mm plug hole, which is not in the line of flow.