Dilution Tanks/Traps, Scientific Plastics
Supplier: Scientific Plastics
W31586-151 W31586-152 W32501-150 W59156-201 W32505-150
89014-044EA 1403.7 CAD
89014-044 89014-046 89014-048 89014-050 89014-052
Dilution Tanks/Traps, Scientific Plastics
Tanks Rectangular Tanks
Lightweight, seamless tanks can be used with water for dilution or with limestone chips for neutralization
Tanks meet ASTM D1248 standards for polyolefin materials.
They are rotationally molded from virgin high-density polyethylene and may be used for continuous service up to 82°C (180°F), or up to 100°C (212°F) in intermittent use. All fittings are made of polyethylene and are welded into all penetrations of tanks.