Nalgene® Three-Way Stopcock, Polypropylene with Teflon® Resin TFE Plug, Thermo Scientific
Supplier: Thermo Fisher Scientific
6470-0002 6470-0004
59097-058CS 756.68 CAD
59097-058 59097-080
Nalgene® Three-Way Stopcock, Polypropylene with Teflon® Resin TFE Plug, Thermo Scientific
Stopcocks
Three-way tubulated outlets arranged in T shape
Flow path directed through any two or all three bores of the stopcock. Non-seizing, autoclavable, and leakproof. Accepts 6.4–7.9mm (1/45/16") tubing.