Nalgene® Stopcocks, Polypropylene with Teflon® Resin TFE Plug, Thermo Scientific
Supplier: Thermo Fisher Scientific
6460-0002 6460-0004
59096-000CS 985.6 CAD
59096-000 59096-055
Nalgene® Stopcocks, Polypropylene with Teflon® Resin TFE Plug, Thermo Scientific
Stopcocks
Serrated tubulation on each end
Accepts 6.4–7.9mm (1/45/16") tubing. Autoclavable.