Nalgene® Nylon Syringe Filters, 4 mm, Thermo Scientific
Supplier: Thermo Fisher Scientific
176-0020 176-0045
28196-050CS 1950.53 CAD
28196-050 28196-052
Nalgene® Nylon Syringe Filters, 4 mm, Thermo Scientific
Filters Syringe Filters
Especially useful in critical applications requiring low contamination levels, such as filtering HPLC samples in aqueous or organic solutions.
  • Non sterile
  • Diameter: 4 mm
  • Low level of extractables
  • Female Luer-lock inlet and male Luer slip outlet
Nylon membrane is inherently hydrophilic and chemically-resistant to a wide range of solvents. Polypropylene housing contains no wetting agents or plasticizers.
Ideal for sterilizing cryoprotectants, such as DMSO.