Whatman™ Cellulose Acetate Membrane Filters
Supplier: Cytiva
10404106 10404006 10404012 10403112 10403012 7000-0004 10404106 10404006 10403012 10404012 10403112 7000-0004 7000-0002 10404026 10404001 10404031 10404114 10404139 10404180 7001-0004 10404044 10404131 10404014 10404126 10404112 10404001 10404014 10404026 10404031 10404044 10404112 10404114 10404126 10404131 10404139 10404180 7000-0002 7001-0004
CA74330-198PK 215 CAD
CA74330-198 CA74330-202 CA74330-204 CA74330-246 CA74330-304 CA89014-302 74330-198 74330-202 74330-304 74330-204 74330-246 89014-302 CA11012-650 CA11012-738 CA11012-740 CA11012-776 CA11012-800 CA11012-904 CA11013-032 CA11013-178 CA11013-808 CA11013-824 CA11013-962 CA11013-966 CA11013-968 10034-932 10034-934 10034-936 10034-938 10034-940 10034-942 10034-944 10034-946 10034-948 10034-950 10034-954 10035-528 10035-740
Whatman™ Cellulose Acetate Membrane Filters
Filters Membrane Filters
Cytiva's Whatman cellulose acetate membrane filters have very low protein binding, which minimizes sample loss when filtering protein-based aqueous samples.
  • Pure cellulose acetate composition, minimizing sample contamination
  • Hydrophilic, for use with aqueous and alcoholic media
  • High solvent resistance, particularly to low molecular weight alcohols
  • High physical strength and a heat resistance up to 180 °C
  • Can be sterilized by most common methods without sacrificing membrane integrity
Due to their high chemical resistance, cellulose acetate membrane filters reliably prepare liquid scintillation cocktail for scintillation counter measurement.
High chemical resistance membranes.
Additionally, the highly heat resistant Whatman cellulose acetate membranes can filter hot liquids and hot gases.