AMCO AEPA-1® Linear Calibration and Instrument Quality Check Kits, GFS Chemicals
Supplier: GFS CHEMICALS INC
85566
85567
85568
85569
85572
66115-048EA
884.75
CAD
66115-048
66115-050
66115-052
66115-054
66115-058
AMCO AEPA-1® Linear Calibration and Instrument Quality Check Kits, GFS Chemicals
Standards
Turbidity Standards
Meets EPA linear calibration recommendations
Formazin kits should be used within 30 days of purchase.
Available in a variety of ranges. Each kit includes a 125mL bottle of each stated NTU value.