Casework, Laminate, Full Height Storage Cabinets, Hinged Panel Doors
Supplier: Case Systems
T0100842436 T0100842448 T0110842424 T0120842424
33501-234EA 3231.32 CAD
33501-234 33501-236 33501-238 33501-240
Casework, Laminate, Full Height Storage Cabinets, Hinged Panel Doors
Furniture Casework
Case Systems plastic laminate furniture combines modular and fixed laminate casework with coordinating hardware, trim, countertops, fixtures and many complementary accessories.
  • Key component of case systems casework is the plastic laminate
  • Case cabinetry gives adaptability necessary in today’s laboratories
A perfect choice for laboratory environments, plastic laminate provides superior resistance to chemicals, scratching, and abrasion. The radiused 3 mm PVC edging on doors and drawers further protects the casework from dents and marring whileMore Product Information