Versapor® Membrane Disc Filters, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
Versapor®
66389 66393 66400 66400 66386 66408 60129 66397 66396 66331 66401 66331 66411 66408 60178 66393 66386 66332 60178 66410 66396 66387 60129 66387 66401 66332 66394 66397 66410 66394 66389 66411
CA28149-716PK 396 CAD
28149-809 CA28149-716 CA28149-638 CA28149-718 CA87003-864 28150-232 CA28149-860 28149-690 28149-770 28149-791 CA87003-816 28149-714 28149-716 28149-638 28149-610 28149-612 28149-810 28149-634 CA28149-809 28149-718 87003-864 CA28149-770 28149-860 CA28149-791 87003-816 CA28149-690 CA28150-232 CA28149-612 CA28149-634 CA28149-810 CA28149-714 CA28149-610
Versapor® Membrane Disc Filters, Cytiva (Formerly Pall Lab)
Filters Membrane Filters
Ideal for prefiltration of difficult-to-filter solutions and serum.
  • For prefiltration
  • Excellent retention
  • Acrylic copolymer
Designed to be used in applications requiring excellent retention properties combined with tensile strengths in excess of 200 bar (3000 psi).
Filter membrane is made of acrylic copolymer on a nonwoven support. Strong, flexible, and nonshedding, both dry and wet. Delivers high flow rates at low differential pressures. Can be used as a prefilter to replace glass fiber filters. UV or gamma sterilizable.
Ordering information: For additional diameters, please contact your Avantor sales representative.