Nylaflo® Membrane Disc Filters
Supplier: Cytiva
66601 66609 66610 66602 66603 66604 66607 66608 66606 66608
CA28140-028PK 306.12 CAD
CA28140-028 CA28140-044 CA28140-143 CA28140-040 CA28140-041 CA28140-042 CA28140-120 CA28140-141 28140-108 28140-141
Nylaflo® Membrane Disc Filters
Filters Membrane Filters
Useful for a wide range of both aqueous and solvent-based applications.
  • Hydrophilic
  • Excellent chemical compatibility
  • Naturally wettable
Hydrophilic nylon filters offer excellent chemical compatibility with esters, bases, and alcohols
Naturally wettable to aqueous solutions. HPLC certified for low levels of UV-absorbing extractables.
Not recommended for acids >1N or halogenated solvents.
Order Now
SPECIFICATIONS
Environmentally Preferable