Acro® 37 TF Vent Devices, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
4464 4465
CA28143-514PK 687.31 CAD
CA28143-514 CA28142-395
Acro® 37 TF Vent Devices, Cytiva (Formerly Pall Lab)
Filters In-Line Filters
Disposable, multipurpose filters for small volume venting, solvent, and gas filtration.
  • Disposable
  • Multipurpose
  • Polypropylene housing
PTFE membrane and polypropylene housing have broad chemical compatibility. Membrane has 0.2 µm pore size and 37 mm diameter with effective filtration area of 7.5 cm². Hose barb connectors are 6.4 to 9.5 mm (¼ to ⅜"). Maximum operating temperature: 100 °C (212 °F). Maximum operating pressure: 4.1 bar (60 psi), bidirectional. Nonsterile.