LABSET® Organic Chemistry Glassware Kit, Kimble Chase
Supplier: Kimble Chase
LABSET®
29057F-125 21805-300 33820-99
89002-134CS 644.28 CAD
89002-134 89002-162 26387-165
LABSET® Organic Chemistry Glassware Kit, Kimble Chase
Microchemistry Kits
Glassware kit contains components made of KIMAX® brand borosilicate glass with full scale, full length [ST] 24/40 joints
  • Heavy duty construction and consistent quality
  • Long, trouble-free life
  • Minimized frequency of chipping or breakage in assembly and use
"The kit contains one each of the listed components. The ground joints have a cylindrical outside wall that provides a firm clamping surface for reduced breakage and greater safety. The separatory funnel has reinforced construction, with extra glass provided around the stopcock. The separatory funnel stopcock barrel features a mirror finish to take full advantage of the natural lubricity of the PTFE stopcock plug. The plugs operate smoothly without grease, and are interchangeable. The case is made of expanded polystyrene. Case dimensions: 48.6Wx35.6Dx12.7Dcm (191/8x14x5"). (Kimble Chase 33820 99)"
Heavy duty construction and consistent quality assure a long, trouble-free life with minimized frequency of chipping or breakage in assembly and use.