VacuCap® and VacuCap® PF Bottle-Top Filters, Sterile, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
4621 4624 4631 4634 TA4622 TA4632 4638 4628 4622 4632
CA28143-316PK 488.61 CAD
CA28143-316 CA28143-317 CA28143-330 CA28143-334 CA28143-338 CA28143-340 CA28139-704 CA28139-706 CA28143-315 CA28143-332
VacuCap® and VacuCap® PF Bottle-Top Filters, Sterile, Cytiva (Formerly Pall Lab)
Filters Bottle Top Filters
For sterilization or clarification of laboratory fluids. Ideal for cell culture and serum-containing media.
  • Ideal for cell culture
  • High flow rate
  • Supor® (hydrophilic polyethersulfone) membrane provides high flow rates
  • Acrylic housing, sterile
Polyester membrane support. Effective filtration area of VacuCap 60 PF filters is 30 cm²; VacuCap 90 PF filters, 60 cm². Filters 28143-340 and 28143-338 come with individual attached tubing for each filter device. All other filters come with one piece of tubing per ten filter devices.
Draws directly from mix vessel and filters directly into the desired container, minimizing transfer steps. Accepts a variety of collection vessels. VacuCap 60 is designed for volumes of 500 ml to 1 L; VacuCap 90 is designed for volumes of 1 to 5 L.
VacuCap PF devices have built-in prefilter that increases throughput of viscous or particulate solutions, such as serum-containing media.
Ordering information: The feedline accessory kit includes tubing and a glass sinker.