KIMBLE® Microscale Threaded Chemistry Kits, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
746000-0000 746000-0005 746200-0000 746200-0005 746210-0000 746210-0005 746220-0000
KT746000-0000CS 1474.57 CAD
KT746000-0000 KT746000-0005 KT746200-0000 KT746200-0005 KT746210-0000 KT746210-0005 KT746220-0000
KIMBLE® Microscale Threaded Chemistry Kits, DWK Life Sciences
Microchemistry Kits
These threaded chemistry kits contain some of the components necessary to perform the experiments described in the following texts: organic laboratory techniques: microscale approach, Pavia, Lampman, Kriz, Engel, Saunders college publishing and microscale organic laboratory, Mayo, Pike, Butcher, John Wiley and Sons.
  • The rugged polyethylene storage case (2.5H×13.5W×10"D) features a die cut foam insert
  • The EKONO-CASE™ is a low-cost corrugated cardboard alternative to plastic
  • Kits fit easily into lab bench drawers
  • Standard taper 14/10 connections
  • High
More Product Information