Robotic Tips for Agilent Systems
Supplier: NEST SCIENTIFIC
352002 352102 352103 352202 352203 352212 352213
77507-938CS 3556.14 CAD
77507-938 77507-942 77507-944 77507-946 77507-948 77507-950 77507-952
Robotic Tips for Agilent Systems
Pipette Tips
NEST robotic tips can be used for research and development and other commonly used applications with needs of high-throughput pipetting in genomics, proteomics, cytomics, immunoassays, metabolomics, biopharmaceuticals.
  • Adopt medical grade polypropylene material, which is manufactured by precision mold, and the material batch is stable
  • DNase-and RNase-free, no heat source contamination
  • Electron beam sterilization
  • Manufacturing facility equipped with class 100000 clean-room, products packing under sterile condition
  • Unique process technology ensures that the inner surface of the tip is smooth and reduces liquid residue
  • Carry out structural drawing and measurement according to the original adapter to ensure that the product has good air tightness and adaptability
  • Online testing equipment is used to control the size and concentricity of products in the production line
Low-retention tips are an excellent choice for transferring viscous liquid, which can effectively reduce the phenomenon of liquid hanging on the wall and make the experiment more accurate.