Pentafluorobenzyl Bromide (PFBBr) Alkylation Reagent

Supplier: Thermo Scientific
TS-58220
10066-388EA 637.76 CAD
10066-388
Pentafluorobenzyl Bromide (PFBBr) Alkylation Reagent
Chromatography Sorbents GC Sorbents
Derivatize carboxyl acids, phenols, and sulfonamides for electron-capture detection (ECD) GC analysis with the Thermo Scientific™ Pentafluorobenzyl Bromide (PFBBr) Alkylation Reagent.

  • Fast reaction times for extraction alkylation technique: ∽20 minutes
  • Analysis of trace organics in asphalt
  • For use with GC
  • Storage at room temperature
  • Hazards: Corrosive
  • Standard components are alpha-Bromo-2,3,4,5,6-pentafluorotoluene
  • Standard concentration: 98 to 100%

Derivatives are highly EC-sensitive, making them useful in low-level determinations of fatty acids. PFBBr has been applied in analyzing trace organics in asphalts and as a fingerprinting technique for identifying asphalt pollutants found in surface waters.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR