Defender™ 3000 Industrial Hybrid Bench Scales
Supplier: Ohaus
30685206 30685207 30685208 30685196 30685201 30685202 30685177 30685178 30685179 30685220 30685221 30685185 30685190 30685195 30685214 30685215
77321-434EA 1821.09 CAD
77321-434 77321-436 77321-438 77321-414 77321-424 77321-426 77321-384 77321-386 77321-388 77321-462 77321-464 77321-400 77321-402 77321-412 77321-450 77321-452
Defender™ 3000 Industrial Hybrid Bench Scales
Balances and Scales
The OHAUS Defender™ 3000 hybrid - i-D33 bench scales are ideal for general weighing, simple counting and checkweighing applications in production, packaging, warehouse, inventory, shipping and receiving areas.
  • Simple, yet rugged, stainless steel design
  • Stainless steel indicator with tactile keys
  • Multicolor backlit LCD display
  • Built in rechargeable battery operation
Results are clearly shown using the large bright backlit LCD display. Display changes color (yellow/green/red) in Checkweighing mode to alert when within the target weight range.