Defender™ 3000 Industrial Floor Scales
Supplier: Ohaus
30837691 30837692 30837693 30837601 30837603 30837604
77321-574EA 3634 CAD
77321-574 77321-576 77321-578 77321-580 77321-582 77321-562 77321-566 77321-568
Defender™ 3000 Industrial Floor Scales
Balances and Scales
Defender™ 3000 Basic industrial floor scale. Painted carbon steel treaded decks with four alloy steel load cells, fixed top plate 5 mm (0.196") thick with structural bracing for stiff construction.
  • Results are clearly shown on the large, bright backlit LCD display
  • In check weighing mode, display changes color (yellow/green/red) to alert users when within the target weight range