Defender™ 3000 Industrial Drum Floor Scales
Supplier: Ohaus
30469909 30469910 30469911 30469912
77434-976EA 3400.82 CAD
77434-976 77434-978 77434-980 77434-982 77434-984
Defender™ 3000 Industrial Drum Floor Scales
Balances and Scales
Convenient portability with floor scale capacity the OHAUS drum scales come in 5 models with platform sizes ranging from 30×30 to 36×36", and capacities ranging from 500 to 2500 lb, giving the flexibility to choose what best suits your weighing needs. The drum scales feature a platform constructed of mild steel with an epoxy paint finish, which means that the scale can stand up to heavy industrial use.
  • A low-profile design with front and back access ramps that make it easy to roll pallet jacks, dollies, carts and drums on to the platform
  • High-density polyurethane wheels provide convenient portability
The Defender 3000 drum scale is a durable portable floor scale designed for industrial applications. It features a mild steel platform with treads, built-in front/back access ramps, 4 load cells (with alloy steel feet and top leveling), and the popular T32M indicator for clear display of weighing results. For rapid portability, the base can be turned onto its side using the stainless steel handles and it can moved around the shop floor using the two built-in wheels.