Defender™ 3000 Industrial Bench Scales
Supplier: Ohaus
30685199 30685200 30685203 30685204 30685205 30685174 30685175 30685176 30685218 30685219 30685183 30685184 30685193 30685194 30685212 30685213
77321-420EA 2418.59 CAD
77321-420 77321-422 77321-428 77321-430 77321-432 77321-378 77321-380 77321-382 77321-458 77321-460 77321-396 77321-398 77321-408 77321-410 77321-446 77321-448
Defender™ 3000 Industrial Bench Scales
Balances and Scales
The OHAUS Defender™ 3000 - i-D33 bench scales are ideal for general weighing, simple counting and checkweighing applications in production, packaging, warehouse, inventory, shipping and receiving areas.
  • Simple, yet rugged tubular-frame base design
  • Indicators with tactile keys
  • Multicolor backlit LCD display
  • AC or battery operation
  • Flexible mounting capabilities
Results are clearly shown using the large, bright backlit LCD display. In checkweighing mode, the display changes color (yellow or green or red) to alert when it is within the target weight range.