Explorer™ Precision Balances
Supplier: Ohaus
30553843 30553844 30553845 30553846 30553847 30553888 30553889 30553890 30553891 30553892 30553893 30553894
77434-998EA 6562.32 CAD
77434-998 77435-000 77435-002 77435-004 77435-006 77435-008 77435-010 77435-012 77435-014 77435-016 77435-018 77435-020
Explorer™ Precision Balances
Balances and Scales
The Explorer series is the flagship precision balance range from OHAUS. Equipped with a weighing cell that's precision-machined from a solid metal block, Explorer balances are extremely accurate and durable, and are ideal for laboratory and industrial environments.
  • Four-level user management with password protection, and un-editable system log plus a high level of configurability make Explorer balances ideal for regulated applications
  • Detachable terminal with a large color touchscreen and programmable IR sensors
  • Frameless draftshield make Explorer precision balances easy and convenient to use
The OHAUS Explorer Series of precision balances combine modern features and design elements to offer unmatched functionality in a line of high-performance balances unlike any other on the market. These smart and intuitive balances simplify even the most complex laboratory measurements.