Precision Balances, PR
Supplier: Ohaus
30430063 30430064 30430065 30430066 30529239 30529067 30529238
77434-838EA 2317.61 CAD
77434-838 77434-840 77434-842 77434-844 77434-996 77434-992 77434-994
Precision Balances, PR
Balances and Scales
Designed for routine weighing applications. The PR is equipped with 3 essential weighing modes, RS-232 connectivity for data transfer and storage and internal calibration (in select models), making it ideal for routine weighing applications.
  • Easy-to-read, bright backlit display and a simple user interface
  • Incredibly easy to operate, with almost no training required
  • The small footprint saves desktop space while providing a large weighing surface
  • Durable construction and features a stainless steel pan to withstand day-to-day use in the workplace
Offering accuracy and repeatability in essential weighing applications in laboratory, industrial and education settings, PR Precision Balances deliver competitive performance at an economical price. Featuring RS232 connectivity for easy communication, and a backlit display and a simple interface for uncomplicated operation.