P-STIK Articulating Arm to Aid Urine Collection
Supplier: Starplex Scientific
P-STIK
CA77530-106CS 644.49 CAD
CA77530-106
P-STIK Articulating Arm to Aid Urine Collection
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Urine Sample Collection/Preparation
P-STIK is a single-use, lightweight device that features an ergonomic articulating arm for the collection of urine specimens in a sanitary and dignified manner. The P-STIK articulating arm is designed for use with 48 mm urine specimen cups, which are sold separately.
  • Acts as an extension of the arm for improved reach when retrieving urine specimens
  • Keeps hands/skin away from the specimen container, ensuring a true midstream urine sample
  • Sanitary and easy-to-use
  • Maintains dignity of the user
  • Individually wrapped with instructions on the wrap
  • Made from recycled plastic
P-STIK protects patient autonomy while freeing up overloaded care providers hands and time. This easy-to-use device is designed with input from healthcare providers with frontline expertise in acute care. For use in long-term care facilities, family physician clinics, maternity clinics, outpatient laboratories, and acute care/emergency departments.
Clients who will benefit from the use of P-STIK include pregnant women, the obese, those with physical disabilities, multiple sclerosis, ALS, muscular dystrophy, the elderly, and parents aiding their children.
Ordering information: Specimen cups must be purchased separately. See accessories tab for available options.