Cricut Mug Press™ Machine
Supplier: D&H
2007804 2009085 2009086
470354-218EA 363.23 CAD
470354-218 470354-288 470354-270
Cricut Mug Press™ Machine
Educational Materials Arts and Crafts Kits and Supplies
Effortlessly create custom printed mugs for fundraisers, spirt swag, and award memorabilia.
  • Cricut Mug Press for 350 to 470 ml (11 to 16 oz.) Infusible Ink compatible mugs
  • Make pro mugs in minutes with easy, one-touch settings
  • Get peel-proof, dishwasher-safe results with Infusible Ink™ materials
  • Thoughtful safety features include auto-off
  • Requires Infusible Ink Transfer Sheets, Pens, or Markers and compatible mug blanks (sold separately)
Give the gift of vibrant, professional-quality, personalized mugs with Cricut Mug Press. Just create a design using Cricut Infusible Ink materials, attach it to the mug, and let the press do the rest. With no manual temperature or pressure settings, it's easy to customize Infusible Ink compatible mugs* with one-of-a-kind art or text. A patent-pending heat plate with zone heating creates excellent, consistent results. Perfect gifts for family, friends, teachers, neighbors, and coworkers have never been this easy. Optimized for Infusible Ink products, this unique mug press is the perfect complement to any Cricut cutting machine. Carefully follow instructions at cricut.com/make-mugs.
Delivery information: Mug Press Includes: Cricut Mug Press™ heat press for mugs, USB cord for activation (product will not function until activated using PC/Mac computer with USB port and internet connection).
Essential Bundle Includes: Ceramic Mug Blank, White - 12oz./340 ml (4 ct)
Infusible Ink™ Pattern, Pink Limeonade, - 4.5\" x 12\"
Infusible Ink™ Pattern, Purple Watersplash - 4.5\" x 12\"
Infusible Ink™ Markers (1.0), Neons (5 ct)
Heat Resistant Tape
*Materials in bundle may be substituted based on availability.
Everything Bundle Includes: Ceramic Mug Blank, White - 12 oz/340 ml (8 ct)
Infusible Ink™ Pattern, Buffalo Check, - 4.5\" x 12\"
Infusible Ink™ Pattern, Green Watercolor - 4.5\" x 12\"
Infusible Ink™ Pattern, Blue Paint Splash - 4.5\" x 12\"
Infusible Ink™, Black - 4.5\" x 12\"
Infusible Ink™, Cherry Red - 4.5\" x 12\"
Infusible Ink™, Ultraviolet - 4.5\" x 12\"
Infusible Ink™ Pens (0.4), Watercolor Splash (5 ct)
Infusible Ink™ Markers (1.0), Neons (5 ct)
Heat Resistant Tape - 2 ct.
*Materials in bundle may be substituted based on availability.