Electrolysis Apparatus, Closed Design
Supplier: GSC INTERNATIONAL SE
N-00009
470356-466EA 264.49 CAD
470356-466
Electrolysis Apparatus, Closed Design
Educational Materials Chemistry Educational Materials Electrochemistry Learning Activities
This Hoffman electrolyzer water chemistry kit lab is a must-have for any chemistry lab.
  • Closed apparatus reduces risk of spills
  • Permanently mounted electrode makes setup a snap
With this Electrolysis apparatus, almost any type of electrolysis experiment can be done safely without compromizing the visual wow-factor. The high-quality, built-in, platinum coated titanium electrodes are resistant to corrosion. The collection tubes are graduated to allow you to measure and to compare the ratio of the two gases that were created during the electrolysis process. Because the apparatus has a small footprint, only a minimal amount of chemicals will be required to perform an electrolysis experiment. The closed design provides a significant level of added safety in the classroom by reducing the potential for spilling chemicals. It is possible to use a chemical solution for several days, as they are contained inside the structure, allowing use over multiple class periods.
Delivery information: Includes: (2) platinum coated titanium electrodes, (2) soft silicon plugs (white), (1) soft silicon plug (blue), (1) cap.