Probes, Very Fine Tip (0.001"), Stainless Steel, Excelta Corp®
Supplier: Excelta
332A 332B 332C 332D 332E 335B 335A
94024-220EA 18.06 CAD
94024-220 94024-222 94024-224 94024-226 94024-228 89411-730 89411-732
Probes, Very Fine Tip (0.001"), Stainless Steel, Excelta Corp®
Dissection Needles
Smallest tip made - great for magnification and very fine work.
  • Extreme fine tips for extreme fine work
  • Available in short or long handle
  • Corrosion resistant
These stainless steel probes have a 1 mil tip. The 335 Series Mini-Probes are 4.75" long. The 332 series are 6.5" long. They feature the smallest, finest tips available on any probe and are ideal for microscopy.
Order Now
SPECIFICATIONS
Environmentally Preferable