Synthware Gas Dispersion Tube, Fritted, Kemtech America
Supplier: Kemtech America
F260810X F260810C F260810M
60003-588EA 64.55 CAD
60003-588 60003-590 60003-592
Synthware Gas Dispersion Tube, Fritted, Kemtech America
Gas Tubes
Tube stem outer diameter is 8 mm. Stem length is 150 mm.
  • Heavy wall borosilicate construction
  • Coarse, medium, and extra coarse frit porosity available
Designed to disperse of scrub gas in liquid.