Cleanroom Wafer Box Tapes, Polyethylene, Ultratape
Supplier: Ultratape Industries
1110WH050P3T 1110WH100P3D 1110WH150P3D 1110WH200P3D 1110WH300P3D 1110BK050P3T 1110BK075P3T 1110BK150P3D 1110BK300P3D 1110BL075P3T 1110BL100P3D 1110BL150P3D 1110BL200P3D 1110BL300P3D 1110CL050P3T 1110CL075P3T 1110CL150P3D 1110CL200P3D 1110OR050P3T 1110OR075P3T 1110OR150P3D 1110OR300P3D 1110CL300P3D 1110RE050P3T 1110RE100P3D 1110RE150P3D 1110RE200P3D 1110RE300P3D 1110GR050P3T 1110GR100P3D 1110GR200P3D 1110YE075P3T 1110YE150P3D 1110YE200P3D 1110YE300P3D 1110BL125P3D
11212-014CS 1766.2 CAD
11212-014 11212-016 11212-018 11212-020 11212-022 11212-024 11212-300 11212-302 11212-306 11212-310 11212-314 11212-316 11212-318 11212-320 11212-322 11212-336 11212-338 11212-342 11212-344 11212-346 11212-348 11212-352 11212-356 11212-358 11212-360 11212-362 11212-364 11212-366 11212-368 11212-370 11212-372 11212-374 11212-376 11212-380 11212-382 11212-386 11212-390 11212-392 11212-394 89203-784
Cleanroom Wafer Box Tapes, Polyethylene, Ultratape
Tapes
This tape is designed specifically for sealing wafer boxes and cleanroom packaging and bag sealing.
  • No residues after prolonged storage
  • Easy release from the roll with low static charge
Tape adheres lightly to plastics, stretches easily around corners, and has low metal content.
Delivery information: Supplied clean and double-bagged.