Synthware Low Form Dewar Flasks, Hemispherical, Kemtech America
Supplier: Kemtech America
F240100 F240250 F240500
60001-184EA 777.15 CAD
60001-184 60001-186 60001-188
Synthware Low Form Dewar Flasks, Hemispherical, Kemtech America
Dewars and Cryogenic Vessels
Flask size shown is the maximum capacity flask that will fit inside the dewar.
  • Heavy wall borosilicate inner construction
  • Aluminum housing
  • Weighted metal base
Hemispherical dewars work great with flasks up to the size listed.