Dewar Vacuum Bottles, Shallow-Form, Pope Scientific
Supplier: Pope Scientific
8110 8120 8130 8325 8335 8345 8315
63884-961EA 770.38 CAD
63884-961 63884-963 63884-965 63884-971 63884-973 63884-975 63884-969
Dewar Vacuum Bottles, Shallow-Form, Pope Scientific
Dewars and Cryogenic Vessels
Shallow-form Dewars permit cooling operations not practical with standard cylindrical or spherical Dewars
Available with hemispherical or flat bottom interiors in protective aluminum base.
These vacuum bottles are made of borosilicate glass.
Applications include rapid freezing and recovery of tissue/organ specimens, insertion of small flasks, agitation in flask with magnetic stirrer, mixing and tamping with stirring rods or pestles. Cooling range is from dry ice/acetone to liquid nitrogen (LN2). Manufactured for use with liquid nitrogen and ideal for use with liquid gases.