Micro Kit for Organic and Environmental Chemistry, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
139479 773900 168353 327342 385772 438764 438782
20762-072CS 94.68 CAD
20762-072 32645-403 32645-582 32645-607 32645-629 32645-709 32645-720
Micro Kit for Organic and Environmental Chemistry, DWK Life Sciences
Microchemistry Kits
This kit provides apparatus for a wide range of organic chemistry.
  • Smooth surface resist contamination
  • Cleaning is simple
  • Can be used interchangeably with any standard taper ground joints
This innovative microscale kit offers many advantages over traditional standard glassware, due to the reduced quantity of chemicals required for experiments
Standard taper Clear-Seal® grease-free joints provide smooth, unground surfaces and can be used interchangeably with any standard taper ground joints. All components are manufactured from Wheaton 33* lowMore Product Information