PCX™ Cleanroom Tape, Polyethylene, Micronova
Supplier: Micronova
PCX-34OR PCX-34YE PCX-1BL PCX-1DB PCX-1DG PCX-1OR PCX-1WH PCX-2DB PCX-2DG PCX-2OR PCX-2WH PCX-2YE PCX-3OR
68399-836CS 2848.58 CAD
68399-836 68399-842 68399-848 68399-858 68399-860 68399-864 68399-866 68399-872 68399-880 68399-912 68399-914 68399-920 68399-924 68399-928 68399-930 68399-944
PCX™ Cleanroom Tape, Polyethylene, Micronova
Tapes
Low density polyethylene tape, low in halogens and leachable chlorides.
  • Special construction lessens the chance of outgassing
  • Chemical resistance and good conformability
Tape can be used for color-coding, sealing, labeling and masking. It can be also used for various other tasks within the cleanroom that require pressure-sensitive qualities. Low-density polyethylene tape is low in halogens and leachable chlorides. Packaged in a cleanroom with special processing to ensure low extractable ions, down to parts per billion.
Ordering information: ForMore Product Information