VWR® Silicone Pump Tubing
Supplier: VWR International
89068-432 89068-434 89068-436 89068-438 89068-440 89068-442 89068-444 89068-446
89068-432EA 472.55 CAD
89068-432 89068-434 89068-436 89068-438 89068-440 89068-442 89068-444 89068-446
VWR® Silicone Pump Tubing
Tubing Transfer Tubing
VWR® Silicone pump tubing is extremely flexible and specially formulated for reliable performance in peristaltic pumps.
  • Extremely flexible low-volatile grade platinum-cured silicone tubing
  • Ideal for pharmaceutical, biomedical, laboratory, food, beverage, and fragrance applications
  • Ultra-smooth interior surface for uniform flow and bacteria resistance
  • Imparts no taste or odor to critical stream
Extremely flexible tubing is specially formulated to withstand repeated compression and release for consistent, reliable performance in peristaltic pumps.
Order Now
SPECIFICATIONS
Environmentally Preferable
Durometer hardness Shore A, 50