Swiveling Squeegee, Vileda Professional-FHP

Supplier: Vileda Professional
119933
89066-050CS 733.59 CAD
89066-050
Swiveling Squeegee, Vileda Professional-FHP
Mops
True cleanroom squeegee swivels 360° for a full range of motion.

  • True cleanroom squeegee swivels 360° for full range of motion
  • Use on floors, walls, and ceilings after applying disinfectant
  • Squeegee spills to the drain in controlled environment
  • Resists autoclave, gamma, and ETO processes
  • Use with all Vileda Professional® handles

Ergonomic handle extends from 99.1–180.3cm (39–71") with a turn of the wrist. Colored tabs (blue, red, green, and yellow) allow for color coding of equipment for specific applications.

Both squeegee and handle are resistant to autoclaving, gamma irradiation, and ETO sterilization.

Use on floors and walls after applying disinfectant. Squeegee spills to the drain in cleanrooms.

Ordering information: Squeegee and handle are sold separately.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR