HelixMark® High-Pressure Braided Reinforced Silicone Tubing, Freudenberg Medical
Supplier: Freudenberg Medical
HelixMark®
60-664-60 60-664-45 60-664-70 60-664-20 60-664-60 60-664-30 60-664-100 60-664-90 60-664-50 60-664-80 60-664-50 60-664-40
CA55566-108EA 3883.69 CAD
CA55566-108 55566-108 55566-118 55566-106 55566-116 55566-104 55566-114 55566-102 55566-112 CA55566-110 55566-100 55566-110
HelixMark® High-Pressure Braided Reinforced Silicone Tubing, Freudenberg Medical
Tubing Transfer Tubing
Tubing is manufactured with a process that incorporates strong polyester braiding between extrusions of platinum-cured silicone.
  • Fully autoclavable and sterilizable
  • Ideal for increased pressure and high temperature applications
  • Minimum burst pressure measured at 20° C (70° F)
  • Material certification and lot traceability available
Durable, reinforced tubing with identifiable blue spiral is suited for increased pressure and/or high-temperature applications. It is manufactured in an FDA registered medical device facility following Good Manufacturing Practices (GMPs) in accordance with the Quality Systems Regulation.
Tubing may be sterilized by autoclave, ethylene oxide, or gamma radiation. Durometer hardness: Shore A, 65.
Certifications: Meets USP Class VI and European Pharmacopoeia 3.1.9 biocompatibility requirements. Certified to meet ISO 13485 quality standards.
Ordering information: Supplied in 25' coil lengths. Tubing is packaged double-bagged in crush-resistant boxes. Material certification and lot traceability are included.