Pharma-50 Tubing, Silicone, DuPont
Supplier: Liveo
4006923 4007677 3100502 4007671 4007679 3100499 3099661
CA47730-784PK 523.34 CAD
CA47730-784 CA47730-786 CA47730-788 CA47730-790 CA47730-792 CA63000-122 CA63000-124 CA63000-126 CA63000-128 CA63000-132 CA63000-134 CA63000-138 CA77776-338 CA89137-864 CA89137-866 CA89137-870 CA89137-872 CA89137-874 CA89137-876 CA89137-880 CA89137-882 CA89137-884 63000-136 89137-868 89137-878 63000-134 63000-126 63000-128 89137-864
Pharma-50 Tubing, Silicone, DuPont
Tubing Transfer Tubing
Premium-grade tubing is ideal for use in pharmaceutical, biotechnology, and veterinary processes for liquid transfer, filling, and moving air or steam
  • Marked for easy identification
  • No phthalates, plasticizers, or other additives
  • No imparted taste or odor
  • Adheres to rigorous acceptance controls for fewer particulates, gels, and bubbles
Offers an excellent extractable profile, flexibility, high resiliency, high tear resistance, and stability over a wide temperature range. Can be sterilized using ethylene oxide, autoclaving, or gamma radiation.
Certifications: Meets or exceeds GMPs, USP Class VI and 3A Sanitary Standard procedures.
Ordering information: Supplied in 50' coil lengths.