Tygon® SPT-3370 IB Food and Beverage Transfer Tubing
Supplier: Saint Gobain Life Sciences
AHJ001718 AHJ001719 AHJ001722
63014-420PK 4247.74 CAD
63014-420 63014-422 63014-424 63014-426 63014-428 63014-430 63014-432
Tygon® SPT-3370 IB Food and Beverage Transfer Tubing
Tubing Transfer Tubing
Natural translucent silicone tubing is ideal for hot fill lines, food handling, stirring vessels, fermenter lines, laboratory, chemical, and resin feeds, viscous and adhesive materials, and D.I water transfer.
  • Braid reinforcement increases pressure resistance
  • Provides flexibility, durability, and chemical and temperature resistance
  • Durometer hardness: Shore A, 70, tensile strength: 1200 psi, autoclavable
  • Platinum-curing process helps ensure the highest level of biocompatibility available
The ultra-smooth inner surface reduces the potential forMore Product Information