Nalgene® 50 Platinum-Cured Silicone Tubing, Thermo Scientific
Supplier: Thermo Fisher Scientific
8060-0040 8060-0060 8060-0070 8060-0120 8060-0130 8060-0190 8060-0200 8060-0260 8060-0310 8060-0140 8060-0100 8060-0080 8060-0050 8060-0020
16211-320CS 399.01 CAD
16211-320 16211-324 16211-326 16211-332 16211-334 16211-338 16211-340 16211-342 16211-344 63014-730 63014-732 63014-734 63014-738 16211-316
Nalgene® 50 Platinum-Cured Silicone Tubing, Thermo Scientific
Tubing Transfer Tubing
Flexible, durable, high-purity tubing is designed for a variety of pump and transfer applications, including those in the pharmaceutical, research laboratory, bioprocess manufacturing, food and beverage, and semiconductor industries.
  • Contains no phthalate plasticizers that can leach out into transported fluid
  • Durometer hardness: Shore A, 50
  • Autoclavable
Tubing resists adhesion of tissue, blood, proteins, and food products while imparting no taste or odor to the fluids. Manufactured with NALGENE® 50 silicone elastomer. Translucent, nontoxic, nonpyrogenic, and nonhemolytic.
Certifications: Complies with USDA, USP Class VI, and 3A standards.
Ordering information: Supplied in 50' coil lengths.
Caution: Note: Not intended for invasive use.