NovaSqueegee™ Cleanroom, Micronova
Supplier: Micronova
RS-12 RS-22 QDNS-12 QDNS-18 QDNS-22 RS-18
68400-524CS 197.78 CAD
68400-524 68400-526 68400-528 68400-530 68400-534 68400-536 68400-538 68400-540 76205-238
NovaSqueegee™ Cleanroom, Micronova
Mops
A stainless steel squeegee tool with a replaceable rubber blade, designed to clean windows, screens, floors, and walls in the cleanroom. Optional PVA blades are fast-drying, low-linting, and low in particulates with a 600% absorbency rate; ideal for electronic and semiconductor cleanrooms.
  • Designed to clean windows, screens, floors, and walls in the cleanroom
  • Made with 304 electropolished stainless steel and Derlin® resin
  • Rubber replacement blades available
  • PVA replacement blades available for absorbency
  • Quick disconnect handle attachment
  • Entire unit is autoclavable
The Micronova Cleanroom NovaSqueegee™ is a stainless steel tool that comes standard with a replaceable rubber blade and features a Quick Disconnect handle attachment for optional use with the SSU series handles for extended reach.