Slim T™ Double Bucket Cart and Wringers, Micronova
Supplier: Micronova
C-7 C-7WH C-7NB
68400-670EA 3033.16 CAD
68400-670 89187-756 10798-896
Slim T™ Double Bucket Cart and Wringers, Micronova
Mops
A simple and effective double bucket cart system that is easy to use, maneuver, and store. Use with a choice of stainless steel, sieve style wringers to meet specific cleaning needs.
  • Simple and effective, double bucket mopping system
  • Designed for use with a variety of flat head and string style mops
  • Crafted from electropolished stainless steel for durability and long life
  • 360° rotation, heavy-duty caster wheels for excellent maneuverability
  • Includes two, 48.3 cm (19") blue and yellow polypropylene buckets
  • 6 gal. (22.7 L) buckets include liter and gallon markings
  • Frame, casters, and buckets are autoclavable
The Slim T™ Double Bucket Cart System is a low profile, electropolished stainless steel frame that secures and easily transports cleaning buckets (included) with excellent maneuverability. C-7: cart system, C-7WH: Entire unit is autoclavable (including wheels), C-7NB: cart only (buckets not included).
The Slim T™ Wringers are electropolished stainless steel, sieve style wringers with a simple design that efficiently eliminates excess liquid, leaving the mop damp enough to effectively clean and sanitize.
Suitable for use with a variety of mops, including SnapMop™, PadMop™, PocketMop™, SlimLine™, SlideMop™, FlatMop™, T-Mop™, Slim T™, and PowerHead™ series mops.