Curtain Cleaner™ Tool and Covers, Micronova
Supplier: Micronova
CCSC-6IR CCH-48 CCSC-2 CCSC-612 CCSC-6
43300-958CS 1340.47 CAD
43300-958 68400-172 68400-176 89133-452 68400-178
Curtain Cleaner™ Tool and Covers, Micronova
Mops
A spring loaded, clamp mop that works to clean and sanitize vinyl strip curtains by clamping at the top and sliding down, effectively cleaning both sides at once. Ideal for critical environments, cleanroom support, and industrial applications.
  • Cover pairs fit over two, 30.5 cm (12") bar clamps
  • Can be used with strong disinfectants
  • Launderable for re-use and autoclavable
Covers available in a choice of fabrics for specialized cleaning:
NovaPoly™ - smooth knit polyester provide excellent disinfectant dispersal.
PolySorb™ - textured polyester for removing residues and dispersing disinfectant.
NovaLite™ - latex bonded with rayon for greater friction resistance and absorbency.
The CurtainCleaner™ tool provides a simple, effective method to clean and sanitize cleanroom strip curtains and softwall enclosures, without the need for a ladder or stool. The durable stainless steel assembly includes an attached, 48" (120 cm), stainless steel handle with precise clamp hand control.
Ordering information: Mop assembly and slipcovers must be ordered separately.